accessibility menu, dialog, popup

UserName

Viewing profile as user:
OSOHN

CAS RN 2295-31-0

IUPAC Name: 1,3-thiazolidine-2,4-dione
Synonyms: 2,4-thiazolidinedione
Molecular Weight (g/mol): 117.12
Molecular Formula: C3H3NO2S
InChi Key: ZOBPZXTWZATXDG-UHFFFAOYSA-N
SMILES: O=C1CSC(=O)N1
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (2)
Physical Form 
  • (6)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (6)
  • (3)