accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 22972-55-0

IUPAC Name: 4-[(1R,6R)-3-methyl-6-(prop-1-en-2-yl)cyclohex-2-en-1-yl]-5-pentylbenzene-1,3-diol
Synonyms: 4-[(1R,6R)-3-methyl-6-(prop-1-en-2-yl)cyclohex-2-en-1-yl]-5-pentylbenzene-1,3-diol
Molecular Weight (g/mol): 314.47
Molecular Formula: C21H30O2
InChi Key: YWEZXUNAYVCODW-RBUKOAKNSA-N
SMILES: CCCCCC1=CC(O)=CC(O)=C1[C@@H]1C=C(C)CC[C@H]1C(C)=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results