accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2298-07-9

IUPAC Name: 4-bromonaphthalen-1-amine
Synonyms: 4-bromonaphthalen-1-amine
Molecular Weight (g/mol): 222.09
Molecular Formula: C10H8BrN
InChi Key: LIUKLAQDPKYBCP-UHFFFAOYSA-N
SMILES: NC1=C2C=CC=CC2=C(Br)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results