accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 22990-34-7

IUPAC Name: 2-(piperidin-4-yl)propan-2-ol
Synonyms: 2-(piperidin-4-yl)propan-2-ol
Molecular Weight (g/mol): 143.23
Molecular Formula: C8H17NO
InChi Key: IIXYTWTZMGUQPT-UHFFFAOYSA-N
SMILES: CC(C)(O)C1CCNCC1
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results