accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 22990-77-8

IUPAC Name: 1-(piperidin-2-yl)methanamine
Synonyms: 1-(piperidin-2-yl)methanamine
Molecular Weight (g/mol): 114.19
Molecular Formula: C6H14N2
InChi Key: RHPBLLCTOLJFPH-UHFFFAOYNA-N
SMILES: NCC1CCCCN1
Percent Purity 
  • (1)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)