accessibility menu, dialog, popup

UserName

Viewing profile as user:
SSOOClNOFFF

CAS RN 229956-98-3

IUPAC Name: 5-[5-(trifluoromethyl)-1,2-oxazol-3-yl]thiophene-2-sulfonyl chloride
Synonyms: 5-[5-(trifluoromethyl)-1,2-oxazol-3-yl]thiophene-2-sulfonyl chloride
Molecular Weight (g/mol): 317.68
Molecular Formula: C8H3ClF3NO3S2
InChi Key: TWGONVLMWWEOFL-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(=NO1)C1=CC=C(S1)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results