accessibility menu, dialog, popup

UserName

Viewing profile as user:
NN

CAS RN 230-07-9

IUPAC Name: 4,7-phenanthroline
Synonyms: 4,7-phenanthroline
Molecular Weight (g/mol): 180.21
Molecular Formula: C12H8N2
InChi Key: DATYUTWESAKQQM-UHFFFAOYSA-N
SMILES: C1=CN=C2C=CC3=NC=CC=C3C2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results