accessibility menu, dialog, popup

UserName

Viewing profile as user:
NN

CAS RN 230-46-6

IUPAC Name: 1,7-phenanthroline
Synonyms: 1,7-phenanthroline
Molecular Weight (g/mol): 180.21
Molecular Formula: C12H8N2
InChi Key: OZKOMUDCMCEDTM-UHFFFAOYSA-N
SMILES: C1=CN=C2C(C=CC3=NC=CC=C23)=C1
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)