CAS RN 23007-85-4
IUPAC Name:
hydrogen 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine chloride
Synonyms:
hydrogen MPTP chloride
Molecular Weight (g/mol):
209.72
Molecular Formula:
C12H16ClN
InChi Key:
KOWJANGMTAZWDT-UHFFFAOYSA-N
SMILES:
[H+].[Cl-].CN1CCC(=CC1)C1=CC=CC=C1
1
–
1
of
1
results
1
1-Methyl-4-phenyl-1,2,3,6-tetrahydropyridine Hydrochloride 98.0+%, TCI America™
CAS: 23007-85-4 Molecular Formula: C12H16ClN Molecular Weight (g/mol): 209.72 MDL Number: MFCD00012790 InChI Key: KOWJANGMTAZWDT-UHFFFAOYSA-N Synonym: MPTP Hydrochloride PubChem CID: 161406 IUPAC Name: hydrogen 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine chloride SMILES: [H+].[Cl-].CN1CCC(=CC1)C1=CC=CC=C1