accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOOOOONOO

CAS RN 23024-29-5

IUPAC Name: bis(2,5-dioxopyrrolidin-1-yl) decanedioate
Synonyms: bis(2,5-dioxopyrrolidin-1-yl) decanedioate
Molecular Weight (g/mol): 396.4
Molecular Formula: C18H24N2O8
InChi Key: XPKIVYVYXKPGBP-UHFFFAOYSA-N
SMILES: O=C(CCCCCCCCC(=O)ON1C(=O)CCC1=O)ON1C(=O)CCC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results