accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2303-16-4

H3CCH3NH3CCH3OSClCl(Z)

CAS RN 2303-16-4

IUPAC Name: N,N-bis(propan-2-yl){[(2Z)-2,3-dichloroprop-2-en-1-yl]sulfanyl}formamide
Synonyms: di-allate
Molecular Weight (g/mol): 270.21
Molecular Formula: C10H17Cl2NOS
InChi Key: SPANOECCGNXGNR-UITAMQMPSA-N
SMILES: CC(C)N(C(C)C)C(=O)SC\C(Cl)=C\Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results