accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3NH3CCH3OSClClCl

CAS RN 2303-17-5

IUPAC Name: N,N-bis(propan-2-yl)[(2,3,3-trichloroprop-2-en-1-yl)sulfanyl]formamide
Synonyms: triallate
Molecular Weight (g/mol): 304.65
Molecular Formula: C10H16Cl3NOS
InChi Key: MWBPRDONLNQCFV-UHFFFAOYSA-N
SMILES: CC(C)N(C(C)C)C(=O)SCC(Cl)=C(Cl)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results