CAS RN 2304-30-5
IUPAC Name:
tetrabutylphosphanium chloride
Synonyms:
tetrabutylphosphonium chloride
Molecular Weight (g/mol):
294.89
Molecular Formula:
C16H36ClP
InChi Key:
IBWGNZVCJVLSHB-UHFFFAOYSA-M
SMILES:
[Cl-].CCCC[P+](CCCC)(CCCC)CCCC
1
–
1
of
1
results
1
Sigma Aldrich 5,10,15,20-Tetrakis(4-Trimethylammoniophenyl)Porphyrin Tetra(P-Toluenesulfonate)
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.