accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2305-05-7

IUPAC Name: 5-octyloxolan-2-one
Synonyms: gamma-dodecalactone
Molecular Weight (g/mol): 198.31
Molecular Formula: C12H22O2
InChi Key: WGPCZPLRVAWXPW-UHFFFAOYNA-N
SMILES: CCCCCCCCC1CCC(=O)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results