accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2305-13-7

IUPAC Name: 4-(3-hydroxypropyl)-2-methoxyphenol
Synonyms: dihydroconiferyl alcohol
Molecular Weight (g/mol): 182.22
Molecular Formula: C10H14O3
InChi Key: MWOMNLDJNQWJMK-UHFFFAOYSA-N
SMILES: COC1=CC(CCCO)=CC=C1O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results