accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2305-26-2

IUPAC Name: cyclohex-4-ene-1,2-dicarboxylic acid
Synonyms: cyclohex-4-ene-1,2-dicarboxylic acid
Molecular Weight (g/mol): 170.16
Molecular Formula: C8H10O4
InChi Key: ILUAAIDVFMVTAU-UHFFFAOYNA-N
SMILES: OC(=O)C1CC=CCC1C(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)