accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NClNOO

CAS RN 23056-40-8

IUPAC Name: 2-chloro-5-methyl-3-nitropyridine
Synonyms: 2-chloro-5-methyl-3-nitropyridine
Molecular Weight (g/mol): 172.57
Molecular Formula: C6H5ClN2O2
InChi Key: LUAJUWOJEFFNFE-UHFFFAOYSA-N
SMILES: CC1=CN=C(Cl)C(=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results