accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNO

CAS RN 230618-24-3

IUPAC Name: 6-(pyrrolidin-1-yl)pyridine-2-carbaldehyde
Synonyms: 6-(pyrrolidin-1-yl)pyridine-2-carbaldehyde
Molecular Weight (g/mol): 176.22
Molecular Formula: C10H12N2O
InChi Key: SZOZYXCTOMKVTL-UHFFFAOYSA-N
SMILES: O=CC1=NC(=CC=C1)N1CCCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results