accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COO

CAS RN 23074-10-4

IUPAC Name: 5-ethylfuran-2-carbaldehyde
Synonyms: 5-ethylfuran-2-carbaldehyde
Molecular Weight (g/mol): 124.14
Molecular Formula: C7H8O2
InChi Key: XADGZBXFWQHBDB-UHFFFAOYSA-N
SMILES: CCC1=CC=C(O1)C=O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)