CAS RN 2309-07-1
IUPAC Name:
methyl (2E)-3-(4-hydroxy-3-methoxyphenyl)prop-2-enoate
Synonyms:
methyl (2E)-3-(4-hydroxy-3-methoxyphenyl)prop-2-enoate
Molecular Weight (g/mol):
208.21
Molecular Formula:
C11H12O4
InChi Key:
AUJXJFHANFIVKH-GQCTYLIASA-N
SMILES:
COC(=O)\C=C\C1=CC=C(O)C(OC)=C1
1
–
1
of
1
results
1
Promotions Available
Methyl 4-hydroxy-3-methoxycinnamate, 99%
Methyl 4-hydroxy-3-methoxycinnamate, 99%
CAS: 2309-07-1 MDL Number: MFCD00017208