accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 23094-96-4

IUPAC Name: 3-bromo-4-methoxybenzene-1-sulfonyl chloride
Synonyms: 3-bromo-4-methoxybenzenesulfonyl chloride
Molecular Weight (g/mol): 285.54
Molecular Formula: C7H6BrClO3S
InChi Key: LTVLATJRJIBHDR-UHFFFAOYSA-N
SMILES: COC1=C(Br)C=C(C=C1)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results