accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 23095-31-0

IUPAC Name: 3,4-dimethoxybenzene-1-sulfonyl chloride
Synonyms: 3,4-dimethoxybenzenesulfonyl chloride
Molecular Weight (g/mol): 236.67
Molecular Formula: C8H9ClO4S
InChi Key: RSJSYCZYQNJQPY-UHFFFAOYSA-N
SMILES: COC1=C(OC)C=C(C=C1)S(Cl)(=O)=O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)