accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3OONNH3CH3CNCH3CH3

CAS RN 23103-98-2

IUPAC Name: 2-(dimethylamino)-5,6-dimethylpyrimidin-4-yl N,N-dimethylcarbamate
Synonyms: pirimicarb pyrimicarb aficida ZZ-aphox pyrimor pirimicarb (substance) pirimor ABOL pirimor granulate caswell no. 359C
Molecular Weight (g/mol): 238.29
Molecular Formula: C11H18N4O2
InChi Key: YFGYUFNIOHWBOB-UHFFFAOYSA-N
SMILES: CN(C)C(=O)OC1=NC(=NC(C)=C1C)N(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results