accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2312-35-8

IUPAC Name: 2-(4-tert-butylphenoxy)cyclohexyl prop-2-yn-1-yl sulfite
Synonyms: comite propargite uniroyal D014 cyclosulfyne caswell no. 130I propargite (iso) epa pesticide chemical code 097601 omait BPPS naugatuck D-014
Molecular Weight (g/mol): 350.47
Molecular Formula: C19H26O4S
InChi Key: ZYHMJXZULPZUED-UHFFFAOYNA-N
SMILES: CC(C)(C)C1=CC=C(OC2CCCCC2OS(=O)OCC#C)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results