accessibility menu, dialog, popup

UserName

Viewing profile as user:
FONHNNICl

CAS RN 231278-20-9

IUPAC Name: N-{3-chloro-4-[(3-fluorophenyl)methoxy]phenyl}-6-iodoquinazolin-4-amine
Synonyms: N-{3-chloro-4-[(3-fluorophenyl)methoxy]phenyl}-6-iodoquinazolin-4-amine
Molecular Weight (g/mol): 505.72
Molecular Formula: C21H14ClFIN3O
InChi Key: UHFPFDMMKYQMLC-UHFFFAOYSA-N
SMILES: FC1=CC=CC(COC2=C(Cl)C=C(NC3=C4C=C(I)C=CC4=NC=N3)C=C2)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)