accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 231291-22-8

IUPAC Name: 6-(trifluoromethyl)pyridine-3-carboxylate
Synonyms: 6-(trifluoromethyl)pyridine-3-carboxylate
Molecular Weight (g/mol): 190.1
Molecular Formula: C7H3F3NO2
InChi Key: JNYLMODTPLSLIF-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=C(N=C1)C(F)(F)F
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)