accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 23132-21-0

IUPAC Name: 2-bromo-3-methyl-5-nitropyridine
Synonyms: 2-bromo-3-methyl-5-nitropyridine
Molecular Weight (g/mol): 217.02
Molecular Formula: C6H5BrN2O2
InChi Key: FZIQHPKXSLHGBZ-UHFFFAOYSA-N
SMILES: CC1=CC(=CN=C1Br)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results