accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCO

CAS RN 23138-50-3

IUPAC Name: 1-ethyl-4-isocyanatobenzene
Synonyms: 1-ethyl-4-isocyanatobenzene
Molecular Weight (g/mol): 147.18
Molecular Formula: C9H9NO
InChi Key: FWPYUSLQCQDLJR-UHFFFAOYSA-N
SMILES: CCC1=CC=C(C=C1)N=C=O
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)