accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOI

CAS RN 2314-37-6

IUPAC Name: 3-iodo-4-methoxybenzaldehyde
Synonyms: 3-iodo-4-methoxybenzaldehyde
Molecular Weight (g/mol): 262.05
Molecular Formula: C8H7IO2
InChi Key: RVWOHBWQJGLXIJ-UHFFFAOYSA-N
SMILES: COC1=C(I)C=C(C=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results