accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 23145-16-6

IUPAC Name: 5-bromo-1-benzofuran-2-carbaldehyde
Synonyms: 5-bromo-1-benzofuran-2-carbaldehyde
Molecular Weight (g/mol): 225.04
Molecular Formula: C9H5BrO2
InChi Key: UPEGFMDITWHVHV-UHFFFAOYSA-N
SMILES: BrC1=CC=C2OC(C=O)=CC2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results