accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 23147-57-1

IUPAC Name: 2,3,5,6-tetramethyl-1,4-dioxane-2,5-diol
Synonyms: 2,3,5,6-tetramethyl-1,4-dioxane-2,5-diol
Molecular Weight (g/mol): 176.21
Molecular Formula: C8H16O4
InChi Key: DFMGATPNJMFDCR-UHFFFAOYNA-N
SMILES: CC1OC(C)(O)C(C)OC1(C)O
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)