accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClH3COONH2OOCH3

CAS RN 23150-65-4

IUPAC Name: 1,5-dimethyl 2-aminopentanedioate hydrochloridyl
Synonyms: 1,5-dimethyl 2-aminopentanedioate hydrochloridyl
Molecular Weight (g/mol): 210.63
Molecular Formula: C7H13ClNO4
InChi Key: BPHCSYSXOKTCMA-UHFFFAOYNA-N
SMILES: [Cl].COC(=O)CCC(N)C(=O)OC
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)