accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 23159-07-1

IUPAC Name: 3-(pyrrolidin-1-yl)propan-1-amine
Synonyms: 3-(pyrrolidin-1-yl)propan-1-amine
Molecular Weight (g/mol): 128.22
Molecular Formula: C7H16N2
InChi Key: VPBWZBGZWHDNKL-UHFFFAOYSA-N
SMILES: NCCCN1CCCC1
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (3)
  • (3)
  • (1)
Boiling Point 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (4)
  • (4)
  • (3)
  • (3)
  • (3)
  • (1)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)