accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 23163-86-2

IUPAC Name: 1-chloro-4-isothiocyanato-2-(trifluoromethyl)benzene
Synonyms: 1-chloro-4-isothiocyanato-2-(trifluoromethyl)benzene
Molecular Weight (g/mol): 237.62
Molecular Formula: C8H3ClF3NS
InChi Key: AHFPRSSHNSGRCU-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=C(Cl)C=CC(=C1)N=C=S
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results