accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 23165-64-2

IUPAC Name: 2-chloro-1-isothiocyanato-4-nitrobenzene
Synonyms: 2-chloro-1-isothiocyanato-4-nitrobenzene
Molecular Weight (g/mol): 214.62
Molecular Formula: C7H3ClN2O2S
InChi Key: JSYNALWWKRNZDX-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(Cl)=C(C=C1)N=C=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results