accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3(S)(R)OHO(R)O(S)OO(R)(R)O(S)O(R)HO(S)HO(S)HO(R)OH

CAS RN 23180-57-6

IUPAC Name: [(1R,2S,3R,5R,6R,8S)-6-hydroxy-8-methyl-3-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-9,10-dioxatetracyclo[4.3.1.0²,⁵.0³,⁸]decan-2-yl]methyl benzoate
Synonyms: paeoniflorin paeony root peoniflorin sulfonate paeonia moutan peoniflorin
Molecular Weight (g/mol): 480.47
Molecular Formula: C23H28O11
InChi Key: YKRGDOXKVOZESV-WRJNSLSBSA-N
SMILES: C[C@@]12C[C@@]3(O)O[C@@H](O1)[C@]1(COC(=O)C4=CC=CC=C4)[C@H]3C[C@]21O[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O
Percent Purity 
  • (1)
  • (1)
Grade 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)