accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONOClCH3H3C

CAS RN 23184-66-9

IUPAC Name: N-(butoxymethyl)-2-chloro-N-(2,6-diethylphenyl)acetamide
Synonyms: machette butachlor machete brand of butachlor epa pesticide chemical code 112301 bilchlor machete C17H26ClNO2 weedout pillarsete butanex
Molecular Weight (g/mol): 311.85
Molecular Formula: C17H26ClNO2
InChi Key: HKPHPIREJKHECO-UHFFFAOYSA-N
SMILES: CCCCOCN(C(=O)CCl)C1=C(CC)C=CC=C1CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results