accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 23193-18-2

IUPAC Name: 1,3-diethyl 2-(2-oxopropyl)propanedioate
Synonyms: 1,3-diethyl 2-(2-oxopropyl)propanedioate
Molecular Weight (g/mol): 216.23
Molecular Formula: C10H16O5
InChi Key: KGZCSZOGMMZHKB-UHFFFAOYSA-N
SMILES: CCOC(=O)C(CC(C)=O)C(=O)OCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results