accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 231953-40-5

IUPAC Name: 1-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]-1,4-diazepane
Synonyms: 1-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]-1,4-diazepane
Molecular Weight (g/mol): 279.69
Molecular Formula: C11H13ClF3N3
InChi Key: PKKLERQDFWUWNM-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(Cl)=C(N=C1)N1CCCNCC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results