accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 23198-55-2

IUPAC Name: 3,5-dimethyl-1-phenyl-1H-pyrazole-4-carbonitrile
Synonyms: 3,5-dimethyl-1-phenylpyrazole-4-carbonitrile
Molecular Weight (g/mol): 197.24
Molecular Formula: C12H11N3
InChi Key: WOFAIVGOIVCZJJ-UHFFFAOYSA-N
SMILES: CC1=NN(C(C)=C1C#N)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results