CAS RN 2321-07-5
Fluorescein, TCI America™
CAS: 2321-07-5 Molecular Formula: C20H12O5 Molecular Weight (g/mol): 332.31 MDL Number: MFCD00005050 InChI Key: GNBHRKFJIUUOQI-UHFFFAOYSA-N Synonym: Acid Yellow 73 PubChem CID: 16850 ChEBI: CHEBI:31624 IUPAC Name: 3',6'-dihydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one SMILES: C1=CC=C2C(=C1)C(=O)OC23C4=C(C=C(C=C4)O)OC5=C3C=CC(=C5)O
Thermo Scientific Chemicals Fluorescein, pure
CAS: 2321-07-5 Molecular Formula: C20H12O5 Molecular Weight (g/mol): 332.31 MDL Number: MFCD00005050 InChI Key: GNBHRKFJIUUOQI-UHFFFAOYSA-N PubChem CID: 16850 ChEBI: CHEBI:31624 IUPAC Name: 3',6'-dihydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one SMILES: C1=CC=C2C(=C1)C(=O)OC23C4=C(C=C(C=C4)O)OC5=C3C=CC(=C5)O
Thermo Scientific Chemicals Fluorescein
CAS: 2321-07-5 Molecular Formula: C20H12O5 Molecular Weight (g/mol): 332.31 MDL Number: MFCD00005050 InChI Key: GNBHRKFJIUUOQI-UHFFFAOYSA-N Synonym: fluorescein,solvent yellow 94,resorcinolphthalein,yellow fluorescein,3,6-fluorandiol,d and c yellow no. 7,fluoresceine,japan yellow 201,c.i. solvent yellow 94,d&c yellow no. 7 PubChem CID: 16850 ChEBI: CHEBI:31624 IUPAC Name: 3',6'-dihydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one SMILES: C1=CC=C2C(=C1)C(=O)OC23C4=C(C=C(C=C4)O)OC5=C3C=CC(=C5)O
Fluorescein, Laser Grade 99%, Thermo Scientific Chemicals
CAS: 2321-07-5 Molecular Formula: C20H12O5 Molecular Weight (g/mol): 332.31 InChI Key: GNBHRKFJIUUOQI-UHFFFAOYSA-N PubChem CID: 16850 ChEBI: CHEBI:31624 IUPAC Name: 3',6'-dihydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one SMILES: C1=CC=C2C(=C1)C(=O)OC23C4=C(C=C(C=C4)O)OC5=C3C=CC(=C5)O
Fluorescein, SPEX CertiPrep™
CAS: 2321-07-5 Molecular Formula: C20H12O5 Molecular Weight (g/mol): 332.31 InChI Key: GNBHRKFJIUUOQI-UHFFFAOYSA-N PubChem CID: 16850 ChEBI: CHEBI:31624 IUPAC Name: 3',6'-dihydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one SMILES: C1=CC=C2C(=C1)C(=O)OC23C4=C(C=C(C=C4)O)OC5=C3C=CC(=C5)O
Fluorescein, Free Acid, Reag. Ph. Eur., Solstice
CAS: 2321-07-5 Molecular Formula: C20H12O5 Molecular Weight (g/mol): 332.31 MDL Number: MFCD00005050 InChI Key: GNBHRKFJIUUOQI-UHFFFAOYSA-N PubChem CID: 16850 ChEBI: CHEBI:31624 IUPAC Name: 3',6'-dihydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one SMILES: C1=CC=C2C(=C1)C(=O)OC23C4=C(C=C(C=C4)O)OC5=C3C=CC(=C5)O