accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 232275-51-3

IUPAC Name: 4-bromo-2,6-dichlorobenzoic acid
Synonyms: 4-bromo-2,6-dichlorobenzoic acid
Molecular Weight (g/mol): 269.9
Molecular Formula: C7H3BrCl2O2
InChi Key: SSWALODYSQLVPK-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(Cl)C=C(Br)C=C1Cl
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results