accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNBrBr

CAS RN 23229-25-6

IUPAC Name: 2,6-dibromopyrazine
Synonyms: 2,6-dibromopyrazine
Molecular Weight (g/mol): 237.88
Molecular Formula: C4H2Br2N2
InChi Key: JXKQTRCEKQCAGH-UHFFFAOYSA-N
SMILES: BrC1=CN=CC(Br)=N1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)