accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 23239-51-2

IUPAC Name: hydrogen 4-(2-{[(1R,2S)-1-hydroxy-1-(4-hydroxyphenyl)propan-2-yl]amino}ethyl)phenol chloride
Synonyms: hydrogen ritodrine chloride
Molecular Weight (g/mol): 323.82
Molecular Formula: C17H22ClNO3
InChi Key: IDLSITKDRVDKRV-XHXSRVRCSA-N
SMILES: [H+].[Cl-].C[C@H](NCCC1=CC=C(O)C=C1)[C@H](O)C1=CC=C(O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results