accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2324-98-3

IUPAC Name: 1,2,4,5-tetrafluoro-3-methoxybenzene
Synonyms: 2,3,5,6-tetrafluoroanisole
Molecular Weight (g/mol): 180.1
Molecular Formula: C7H4F4O
InChi Key: AXCOCGJDERQVDK-UHFFFAOYSA-N
SMILES: COC1=C(F)C(F)=CC(F)=C1F
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)