CAS RN 23268-00-0
IUPAC Name:
dihydrogen hydrazine dibromide
Synonyms:
dihydrogen hydrazine dibromide
Molecular Weight (g/mol):
193.87
Molecular Formula:
Br2H6N2
InChi Key:
MEBDASBFCISMGU-UHFFFAOYSA-N
SMILES:
[H+].[H+].[Br-].[Br-].NN
1
–
1
of
1
results
1
Hydrazine Dihydrobromide Hydrate 98.0+%, TCI America™
CAS: 23268-00-0 Molecular Formula: Br2H6N2 Molecular Weight (g/mol): 193.87 MDL Number: MFCD00070601 InChI Key: MEBDASBFCISMGU-UHFFFAOYSA-N Synonym: Hydrazinium Dibromide PubChem CID: 168095 IUPAC Name: dihydrogen hydrazine dibromide SMILES: [H+].[H+].[Br-].[Br-].NN