accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 23271-47-8

IUPAC Name: 1-propylpyridin-1-ium chloride
Synonyms: 1-propylpyridin-1-ium chloride
Molecular Weight (g/mol): 157.64
Molecular Formula: C8H12ClN
InChi Key: IBOIUWAYPMADRC-UHFFFAOYSA-M
SMILES: [Cl-].CCC[N+]1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results