CAS RN 23287-26-5
IUPAC Name:
methyl 2-hydroxy-3-methylbenzoate
Synonyms:
methyl 2-hydroxy-3-methylbenzoate
Molecular Weight (g/mol):
166.18
Molecular Formula:
C9H10O3
InChi Key:
SUHLUMKZPUMAFP-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC=CC(C)=C1O
1
–
1
of
1
results
1
Promotions Available
Methyl 2-hydroxy-3-methylbenzoate, 97%
Methyl 2-hydroxy-3-methylbenzoate, 97%
CAS: 23287-26-5 Molecular Formula: C9H10O3 Molecular Weight (g/mol): 166.18 MDL Number: MFCD00020051 InChI Key: SUHLUMKZPUMAFP-UHFFFAOYSA-N Synonym: methyl 3-methylsalicylate,methyl o-cresotinate,benzoic acid, 2-hydroxy-3-methyl-, methyl ester,levegal pt,2-hydroxy-3-methylbenzoic acid methyl ester,2,3-cresotic acid, methyl ester,methyl-3-methylsalicylate,methyl 2-hydroxy-3-methyl-benzoate,methyl o-cresotate,3-methylsalicylic acid methyl ester PubChem CID: 31835 IUPAC Name: methyl 2-hydroxy-3-methylbenzoate SMILES: COC(=O)C1=CC=CC(C)=C1O