accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 23350-58-5

IUPAC Name: (2E)-but-2-enamide
Synonyms: 2-butenamide (9CI)
Molecular Weight (g/mol): 85.11
Molecular Formula: C4H7NO
InChi Key: NQQRXZOPZBKCNF-NSCUHMNNSA-N
SMILES: C\C=C\C(N)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results