accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 23351-91-9

IUPAC Name: 5-bromobenzene-1,3-dicarboxylate
Synonyms: 5-bromobenzene-1,3-dicarboxylate
Molecular Weight (g/mol): 243.01
Molecular Formula: C8H3BrO4
InChi Key: JATKASGNRMGFSW-UHFFFAOYSA-L
SMILES: [O-]C(=O)C1=CC(=CC(Br)=C1)C([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)